Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Food Standards
Reference standards for food analysis, FAME mixtures
Reference standards for food analysis, FAME mixtures
Reference standards for food analysis, FAME mixtures
Catalog # ZZ49LA90-1100
Supplier:
VWR International
CAS Number:
Expand image
Reference standards for food analysis, FAME mixtures
Catalog # ZZ49LA90-1100
Supplier:
VWR International
CAS Number:
Specifications
Pk:
10 mg
Description:
Me 100
Specifications
About this item
Product Details
Product Details