Preparative HPLC columns, Ultra IBD
Supplier: Restek
Certificates
About this item
Ultra IBD is a polar-embedded column that acts as a strong hydrogen bonder and may be the most versatile column available today. It is compatible with 100% aqueous mobile phases and can be used under reversed-phase or HILIC conditions to retain very polar, ionic compounds in highly organic mobile phases.
- With a unique polar group, this column is very retentive and selective for acids
- Provides symmetrical peak shape for strong bases
Specifications
- Catalog No:
- REST9175558
- REST9175557
- REST9175579
- REST9175519
- REST9175578
- REST9175577
- REST9175518
- REST9175517
- REST9175569
- REST9175568
- REST9175567
- REST9175559
- Carbon load:
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- Endcapped:
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- N/A
- pH range:
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- Temperature limit:
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- USP code:
- L68
- L68
- L68
- L68
- L68
- L68
- L68
- L68
- L68
- L68
- L68
- L68