GC columns, Rt®-βDEXsa (fused silica)
Supplier: Restek
Certificates
About this item
The Rt®-βDEXsa columns have a unique selectivity for esters, lactones, and other fruit flavor components.
Frequently Bought Together
The Rt®-βDEXsa columns have a unique selectivity for esters, lactones, and other fruit flavor components.