Anti-Brain1/POU3F3 Rabbit Polyclonal Antibody
BOSSBS-11947R
: Bioss
- Antibody type:Primary
- Antigen symbol:Brain1/POU3F3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:Brain-specic homeobox/POU domain protein 1|Brain1|RHS 1|OTF-8|Octamer-binding protein 8|BRN 1|Brain-1|RHS1|PO3 F3|PO3F3|class 3|Skin 1|Skin1|Octamer-binding transcription factor 8|transcription factor 3.|Rhs 2|PO3F3_HUMAN|BRN1|transcription factor 3|POU3F 3|Rhs2|Pou3f3|Brain specic homeobox/POU domain protein 1|OTF8|POU domain|OTF 8|PO3F 3|POU3 F3|Brn-1|POU class 3 homeobox 3|Oct-8|Brain 1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A
- Size:100 μg
- Pk:100 µl
Type: Primary
Antigen: Brain1/POU3F3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat