Anti-FOG2 Rabbit Polyclonal Antibody
Catalog # BOSSBS-11942R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Zinc Finger Protein, FOG Family Member 2
- Antigen symbol:FOG2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:Zinc finger protein ZFPM2.|ZFPM2|Friend of GATA 2|FOG-2|Zinc finger protein multitype 2 |FOG2_HUMAN|hFOG-2 |Zinc finger protein 89B|Friend of GATA2 |Friend of GATA protein 2|Zinc finger protein M2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: FOG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat