Anti-FCGR1A Rabbit Polyclonal Antibody
BOSSBS-3511R
: Bioss
- Antibody type:Primary
- Antigen name:Fc fragment of IgG, high affinity Ia, receptor (CD64)
- Antigen symbol:FCGR1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:2209
- Antigen synonyms:FCRI|IGFR1|CD64|CD64A
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Immunogen:100-150/374
- Purification:Purified by Protein A
- Size:100 μL
- Pk:100 µl
Type: Primary
Antigen: FCGR1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat