UHPLC columns, Pinnacle® DB PAH UHPLC
Supplier: Restek
Certificates
About this item
Base-deactivated spherical silica that is optimised for UHPLC stability. Designed to resolve complex mixtures of polycyclic aromatic hydrocarbons (PAHs). Complete resolution of all 16 EPA 610 PAHs, plus two other routinely analyzed PAH compounds, in less than five minutes to greatly reduce run times and increase sample throughput.
Specifications
- Catalog No:
- REST9470212
- REST9470232
- REST9470252
- Endcapped:
- N/A
- N/A
- N/A
- pH range:
- 2,5 - 8
- 2,5 - 8
- 2,5 - 8
- Temperature limit:
- 80 °C
- 80 °C
- 80 °C