Mouse E-selectin ELISA kit
BIRBORB99236-96
: Biorbyt
:
- Pk:96
- Host:
- Primary antibody reactivity:Mouse
- Description:Mouse E-Selectin ELISA Kit
- Environmentally Preferable:
- Detection range:62,5 - 4000 pg/ml
- Sensitivity:<10 pg/ml
Mouse E-Selectin ELISA Kit for the quantitative detection of mouse soluble E-Selectin in cell culture supernates, serum and plasma (heparin, citrate).
- No detectable cross-reactivity with other relevant proteins