Visual comparator test kits, chlorine (total), model CN-21P
Catalog # 705-0136
Supplier: Hach
CAS Number:
Specifications
- Pk:1 KIT
- Description:Chlorine (total), CN-21P
- Tests per kit:100
- Range:10 - 200 mg/l
Specifications
About this item
Qualitative results using traditional drop count titrimetric methods.
- Portable - ideal for field testing
- Simple fast method
- Kits contain all required reagents and apparatus in a rugged carrying case for 100 tests
Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.