Cell culture media, liquid, Dulbecco's Modified Eagle's Medium (DMEM), BioWhittaker™
Catalog # 733-1729
Supplier: LONZA
CAS Number:
Specifications
- Media Format:Liquid
- Pk:500 ml
- Description:DMEM with 4,5 g/L glucose, without L-glutamine (hybridoma fusion screened)
- Seq:12
- Environmentally Preferable:
Specifications
About this item
Chemically defined liquid media is used to provide nutrients for cell culture growth in research, diagnostic and manufacturing applications. All BioWhittaker™ classical media products are manufactured in accordance with cGMP regulations.
DMEM is used in a wide range of mammalian cell culture applications. The high glucose version is well suited to high density suspension culture. The low glucose formula is used for adherent dependent cells.