Chemical resistant cleanroom overalls, Kimtech™ A8
Supplier: KIMBERLY CLARK
Certificates
About this item
These overalls made of ultra light breathable SMS fabric provide Type 6 limited chemical splash protection and Type 5 particle protection.
- Low lint fabric, Helmke Drum Category III
- 94% bacterial filtration efficiency
- Unhooded design with high neck collar
- Elastic cuffs and built-in thumb loops to reduce skin exposure
- Fabric is anti-static treated
- No label on the outer surface
Ideal for use in non sterile pharmaceutical manufacturing, in bio-medical research, pharmaceutical compounding and biotechnology. Suitable for use in ISO 7 & 8/Grade C & D cleanrooms.
Weight: 45 g/m²
Certifications: PPE Cat III (EU) 2016/425, Type 5/6, EN1073-2, EN1149-5
Packaging: One large bag of 25 overalls individually packed per case.
Specifications
- Catalog No:
- 113-8622
- 113-8627
- 113-8625
- 113-8621
- 113-8626
- 113-8623
- 113-8624
- Model:
- A8
- A8
- A8
- A8
- A8
- A8
- A8
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Anti-static:
- +
- +
- +
- +
- +
- +
- +
- Cleanroom class:
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- Garment application:
- Pharmaceutical, Laboratory
- Pharmaceutical, Laboratory
- Pharmaceutical, Laboratory
- Pharmaceutical, Laboratory
- Pharmaceutical, Laboratory
- Pharmaceutical, Laboratory
- Pharmaceutical, Laboratory
- Bacterial filtration efficiency:
- 94%
- 94%
- 94%
- 94%
- 94%
- 94%
- 94%
- Particle shedding (Helmke Drum):
- Level III
- Level III
- Level III
- Level III
- Level III
- Level III
- Level III
- Weight:
- 45 g/m²
- 45 g/m²
- 45 g/m²
- 45 g/m²
- 45 g/m²
- 45 g/m²
- 45 g/m²