ContactUsLinkComponent
Biopsy cassettes, fixed lid
Biopsy cassettes, fixed lid
Catalog # KLIN510-03S
Supplier:  KLINIPATH
Biopsy cassettes, fixed lid
Catalog # KLIN510-03S
Supplier:  KLINIPATH
Specifications
  • Colour:
    Blue
  • Seq:
    03
  • Pk:
    1.200
Specifications
About this item
Biopsy cassettes with fixed lid.
  • Cassettes maintain their stability, rigidness and shape during processing of tissues
  • Available in different colours
Packaging: Taped stacks contain 40 cassettes which are specially packed for Sakura® / Leica® cassette printers.