Orders LinkContactUsLinkComponent
Embedding cassettes, without lid
Embedding cassettes, without lid
Pink
Catalog # KLIN310-04T
Supplier:   KLINIPATH
Embedding cassettes, without lid
Catalog # KLIN310-04T
Supplier:  KLINIPATH
Embedding cassettes, without lid
Embedding cassettes, without lid
Pink
Price
314,00 €
Log in for availability
1 * 3.200 items
Product Details & Documents
Every cassette maintain its stability, rigidness and shape during processing of tissues.
Packaging: Hoppers contain 75 standard cassettes and are specially packed for Thermo Fisher® cassette printers. Taped stacks contain 40 cassettes and are specially packed for Sakura® / Leica® cassette printers.
Specifications
  • Colour:
    Pink
  • Seq:
    28