Orders LinkContactUsLinkComponent
First aid kit, large
First aid kit, large
First aid kit, large, durable EVA case (green/yellow), with handle, 310×80×247 mm
Catalog # 118-0395
Supplier:   ORKLA CARE
First aid kit, large
Catalog # 118-0395
Supplier:  ORKLA CARE
First aid kit, large
First aid kit, large
First aid kit, large, durable EVA case (green/yellow), with handle, 310×80×247 mm
Price
146,00 €
Log in for availability
1 * 1 items
Product Details & Documents
This portable first aid kit contains first aid products to be used in emergency situations (bleeding, breathing, burns, sprains, circulatory shock). Ideal for people who are often on the go and as a complement to a wall-mounted first aid station.
  • Strong and durable case is made from a rubber-like material (EVA) with a matte finish
  • Durable rubber handle makes it easy to carry the kit around
  • Products are packed in transparent plastic pockets: Easy to find the product that is needed
  • All products are protected against dust, dirt and moisture
  • Products come with clear, self-explanatory instructions
  • Enough space for additional products
Contents:
3× Cederroth 4-in-1 Bloodstoppers
2× Cederroth 4-in-1 mini Bloodstoppers
20× Salvequick plasters
10× Salvequick wound cleansers
1× Soft foam bandage (6×40 cm)
1× Breathing mask
2× Safety hand cleanser
2× Pairs of gloves
1× Rescue blanket
2× Elastic bandages
2× Metallised dressings (20×40 cm)
1× Fixing nets
1× Cederroth burn gel dressing
1× Triangular bandage
1× Pair of scissors
1× First aid instructions
Shelf life: 5 years from production date for each component
Specifications
  • Description:
    First aid kit, large, durable EVA case (green/yellow), with handle
  • Dimensions:
    310×80×247 mm