Multi-position magnetic stirrers, Variomag Biosystem Direct
Catalog # 442-3119
Supplier: THERMO H + P LABORTECHNIK
Specifications
- No. of stirring positions:1
- Description:Biosystem Direct, UK-plug, 1 position
- Seq:01
- Environmentally Preferable:
- Model:Biosystem Direct
- Speed range:5 - 120 min⁻¹
- W×D×H:180×180×60 mm
- Max. stirring capacity per stirrer H₂O:5000 ml
- Plate dimensions:180×180 mm
- Plate material:Stainless steel
- Power input:1 W
- Weight:3,4 kg
- Pk:1
- Stirring capacity:5000 ml
- For:4 vessels
Specifications
About this item
Slow speed stirrers with low heat output and gentle stir acceleration/deceleration mainly designed for demanding cell culture application and thermally sensitive operations.
- Reliability and safety
- Heatless operation protects heat-sensitive cell culture
- Stainless-steel housing is easy to clean and resists aggressive media
- Gradual low-shear acceleration on start-up
- Operating conditions are −10 to +40 °C at 95 % RH
Certifications: Class IP6 protection and compliance with CE marked.
Delivery: Supplied with 100 to 240 V units which includes a cord set with various plugs.