Modular concentrators, digital series, SpeedVac™
Supplier: THERMO LABSYSTEMS LIFE SCIENCE
Certificates
About this item
These SpeedVac™ concentrators are ideal for conducting basic molecular biology applications and organic chemistry applications.
- Automatic bleeder valve for un-attended start/stop
- Digital display of parameters for easy, reproducible selection
- Independent timers for both heat and run
- PTFE-coated chamber for superior chemical resistance and easy cleaning
- Modular: Can be combined with an appropriate cold trap, vacuum pump or chemical trap for a complete concentration/drying system
Model SPD111V:
- Basic molecular biology applications with increased microplate capacity
- Simple display and programming
Model SPD121P:
- Enhanced evaporation model, organic compatible
- Radient glass cover to accelerate evaporation and reduce drying time
- Vacuum end point mode terminates run when preset vacuum level is reached
- Digital display of vacuum, temperature and time section
Ordering information: Rotors need to be ordered separately.
Frequently Bought Together