Rapid phosphatase test, lactognost
Catalog # GEAG06.2010
Supplier: Gerber Instruments AG
CAS Number:
Specifications
- Pk:1 KIT
- Description:Lactognost Rapid Phosphatase Kit, with reagents for 100 tests, dosing spoon, colour comparative table and instructions for use
Specifications
About this item
For the detection of alkaline phosphatase in milk, whey, cream and butter.
The test can be used to determine sufficient HTST treatment of milk and related products, and to detect raw milk in pasteurised milk.