ContactUsLinkComponent
Cryogenic vial grippers, Corning®
Cryogenic vial grippers, Corning®
Catalog # 479-1846
Supplier:  Corning
undefined
Cryogenic vial grippers, Corning®
Catalog # 479-1846
Supplier:  Corning
Specifications
  • Description:
    Cryogenic vial grippers, assorted colours
  • Seq:
    01
  • Pk:
    5
Specifications
About this item
These cryogenic vial grippers feature a unique design to grasp internal or external-thread cryogenic vials.
They enable to easily sort or move vials while maintaining sterility and protecting fingers from frozen vials, dry ice and liquid nitrogen.