Pierce™ Alkaline Phosphatase, Biotinylated
Catalog # PIER29339
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 mg
- Description:Alkaline phosphatase, biotinylated
Specifications
About this item
Biotinylated alkaline phosphatase is most commonly used in immunohistochemistry (IHC) to amplify the signal of biotinylated primary antibodies using the ABC staining method.