Neuraminidase
Catalog # J63259.MA
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:10 mg
- CAS No.:9001-67-6
- Storage conditions:–30...–10 °C
- Form:Lyophilized powder
- MDL No.:MFCD00131711
- Enzyme name:Neuraminidase
- Solubility:Soluble in water or 10 mM phosphate buffer, pH 6.0
Specifications
About this item
Applications: A hydrolytic enzyme that removes sialic acid from mucoproteins