Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Timers
Digital Timers
Digital bench top timer
Digital bench top timer
Digital bench top timer
Catalog # LPIT291137
Supplier:
LP ITALIANA
Expand image
Digital bench top timer
Catalog # LPIT291137
Supplier:
LP ITALIANA
Specifications
Colour:
White
Description:
Bench top timer, electronic, ABS casing
W×D×H:
65×50×23 mm
Pk:
1
Specifications
About this item
Product Details
Product Details