ContactUsLinkComponent
VWR®, Staining Rack
  VWRU15153-821
undefined
VWR®, Staining Rack
  VWRU15153-821
  • Description:
    Micro slide staining rack
  • Pk:
    1
Stainless steel.
  • Triangular slide with a tapered top
  • Contact the slides only in a narrow zone
  • Simple and quickly adjustable
  • Fit sinks up to 534 mm inside dimensions
Staining rack is designed for use across laboratory sinks of various sizes.