ContactUsLinkComponent
VWR®, Desiccator Plates, Large
  VWRU89038-054
undefined
VWR®, Desiccator Plates, Large
  VWRU89038-054
  • Ø:
    230 mm
  • Type:
    Porcelain
  • Pk:
    1
Porcelain, glazed on one side.
  • Features eight holes with 30 mm Ø, does not have a central hole
  • Without feet