VWR® HistoBond® VistaVision™, Adhesion slides
Catalog # VWRU16004-406
Supplier: VWR Collection
Specifications
- W×D:75×25 mm
- Colour:White
- Description:Adhesion slides, Histobond®
- Seq:01
- Thickness:1 mm
- Environmentally Preferable:
- Pk:72
Specifications
About this item
These slides surfaces are loaded with a permanent positive charge for in situ hybridisation.
- Colour-coded ends facilitate sorting and identification
- Uniform size and thickness
- Firmly anchors all types of sections
The treatment prevents the loss of tissue sections or cells to frequent washing and eliminates the disturbing background effect caused by traditional adhesives.