Specifications
- Antibody type:Primary
- Antigen name:B melanoma antigen 3
- Antigen symbol:BAGE3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Form:liquid
- Antigen synonyms:B melanoma antigen 3|BAGE3|Cancer/testis antigen 2.3|Cancer/testis antigen family 2 member 3|CT2.3.|B melanoma antigen family member 3|BAGE3_HUMAN
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Unknown. Candidate gene encoding tumor antigens.Members of the BAGE gene family encode antigens that are recognized by cytotoxic T lymphocytes and are also known as CT (cancer/testis) antigens. Generated by juxtacentromeric shuffling of the MLL3 gene, the ancestral BAGE gene was expanded by acrocentric exchanges and/or juxtacentromeric movements.Generally, BAGE proteins are silent in all normal tissues with the exception of testis. BAGE2 and BAGE 3 (B melanoma antigen 2 and 3, respectively), also known as Cancer/testis antigen 2.2 and 2.3 (respectively), are 109 amino acid secreted proteins that are expressed in 22% of melanomas, lung and bladder carcinomas, and are also expressed in normal testis tissue. Like the genes encoding MAGE proteins, BAGE genes are most likely silenced by DNA methylation and/or chromatin compaction in normal tissues other than testis.
IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: BAGE3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human