Anti-FBXO4 Rabbit Polyclonal Antibody
Catalog # BOSSBS-9087R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:F-box protein 4
- Antigen symbol:FBXO4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:26272
- Antigen synonyms:DKFZp547N213|F box protein 4|FLJ10141.|F box protein Fbx4|FBX4|FBXO 4|F box only protein 4
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
FBXO4 is a substrate recognition component of a SCF (SKP1-CUL1-F-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins. It probably recognizes and binds to phosphorylated target proteins. It recognizes TERF1 and promotes its ubiquitination together with UBE2D1.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: FBXO4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat