Anti-NET1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-6167R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Neuroepithelial Cell Transforming 1
- Antigen symbol:NET1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:10276
- Antigen synonyms:ARHGEF8|NET1A|neuroepithelial cell transforming gene 1|NET1|Rho guanine nucleotide exchange factor (GEF) 8|p65 Net1 proto-oncogene protein|guanine nucleotide regulatory protein (oncogene)|neuroepithelioma transforming gene 1|small GTP-binding protein regulator
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
This gene is part of the family of Rho guanine nucleotide exchange factors. Members of this family activate Rho proteins by catalyzing the exchange of GDP for GTP. The protein encoded by this gene interacts with RhoA within the cell nucleus and may play a role in repairing DNA damage after ionizing radiation. Pseudogenes of this gene are located on the long arms of chromosomes 1, 7 and 18. Alternative splicing results in multiple transcript variants that encode different protein isoforms.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: NET1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat