Orders LinkContactUsLinkComponent
Containers, rectangular
Containers, rectangular
Supplier:   SEMADENI
Containers, rectangular
Supplier:  SEMADENI
Certificates
Containers, rectangular
4 Options Available Below
Product Details & Documents
HDPE or PP, white, stackable.
These are ideal containers with side handles and edge for gripping to handle solid samples. Containers over 225 mm height are not suitable for liquids.
Product Family Options
Product Information
  • Capacity
    W×D×H
    Colour
    Material
    Availability
    Price
  • 8,5 l
    570×350×70 mm
    White
    PP
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 36,90 €
    36,90 €
    1 * 1 items
       36,90 €
  • 12,5 l
    395×300×165 mm
    White
    HDPE
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 47,80 €
    47,80 €
    1 * 1 items
       47,80 €
  • 23 l
    460×330×205 mm
    White
    HDPE
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 48,20 €
    48,20 €
    1 * 1 items
       48,20 €
  • 29 l
    670×450×125 mm
    White
    HDPE
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 66,20 €
    66,20 €
    1 * 1 items
       66,20 €