Precision density hydrometers, L50
Supplier: Ludwig Schneider GmbH & CO. KG
Certificates
About this item
These precision density hydrometers (with or without thermometers) are suitable for calibration.
Error limit: ±0,0005 g/ml
Certifications: DIN 12791
Packaging: Each packed in a square plastic sleeve.
Specifications
- Catalog No:
- SCNL3132025
- SCNL3132003
- SCNL3132024
- SCNL3132002
- SCNL3132023
- SCNL3132001
- SCNL3132022
- SCNL3132021
- SCNL3132020
- SCNL3132019
- SCNL3132008
- SCNL3132007
- SCNL3132028
- SCNL3132006
- SCNL3132005
- SCNL3132027
- SCNL3132026
- SCNL3132004
- SCNL3142016S
- SCNL3142017S
- SCNL3142018S
- SCNL3142019S
- SCNL3132014
- SCNL3132013
- SCNL3132012
- SCNL3132011
- SCNL3132010
- SCNL3132009
- SCNL3132018
- SCNL3132017
- SCNL3132016
- SCNL3132015
- Reference temperature:
- 20 °C
- 20 °C
- 20 °C
- 20 °C
- 20 °C
- 20 °C
- 20 °C
- 20 °C
- 20 °C
- 20 °C
- 20 °C
- 20 °C
- 20 °C
- 20 °C
- 20 °C
- 20 °C
- 20 °C
- 20 °C
- 20 °C
- 20 °C
- 20 °C
- 20 °C
- 20 °C
- 20 °C
- 20 °C
- 20 °C
- 20 °C
- 20 °C