Orders LinkContactUsLinkComponent
Dust test kit
Dust test kit
Dust test kit
Catalog # SIMESP3200
Supplier:   Industrial Physics
Dust test kit
Catalog # SIMESP3200
Supplier:  Industrial Physics
Dust test kit
Dust test kit
Dust test kit
Price
147,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Dust test kit allows assessment of the quantity and size of dust particles on surfaces prepared for painting.
  • Report forms for five test results to be filled in pre-defined areas
  • Comparator display board to assess dust quantity and particle size
  • Sample transfer sheet in resistant and transparent plastic with pre-printed positions
Used either as a pass or fail test or event as a permanent record of the presence of dust.
Certifications: ISO 8502-3
Delivery information: Supplied with two AA batteries, illuminated magnifier, adhesive tape, pair of scissors, pictorial manual, comparator board, transfer sheet and report forms.
Documents
Specifications
  • Description:
    Dust test kit
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Display board (×2)
    Display board (×2)
    SIMESP3220
    Log in for availability
    1 * 1 items - 22,20 €
Product Information
  • Catalog #
    Availability
    Price
  • Standardized tape
    Standardized tape
    SIMESP3209
    Log in for availability
    1 * 1 items - 40,70 €
Product Information
  • Catalog #
    Availability
    Price
  • Report forms
    Report forms
    SIMESP3222
    Log in for availability
    1 * 125 items - 35,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample transfer sheet (×10)
    Sample transfer sheet (×10)
    SIMESP3221
    Log in for availability
    1 * 1 items - 225,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Spring loaded roller in titanium anodised aluminum
    Spring loaded roller in titanium anodised aluminum
    SIMESP3600
    Log in for availability
    1 * 1 items - 528,00 €