HPLC columns, Allure® PFP propyl (USP L43)
Supplier: Restek
Certificates
About this item
PFP propyl columns have a pentafluorophenyl phase with a propyl spacer. It is highly retentive for basic analytes and phase for separating nucleosides, nucleotides, purines, pyrimidines, halogenated compounds, β-blockers and tricyclic antidepressants.
Specifications
- Catalog No:
- REST9169565
- REST9169572
- REST9169555
- REST9169535
- REST9169511
- REST9169561
- REST9169532
- REST9169562
- REST916950210
- REST916950212
- REST9169575
- REST916950220
- REST9169552
- REST916950222
- REST9169515
- REST9169552-700
- REST9169512
- Carbon load:
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- 17%
- Endcapped:
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes
- Yes
- Yes, Yes
- Yes
- Yes, Yes
- Yes
- Yes, Yes
- Yes
- Yes, Yes
- pH range:
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- 2,5 - 7,5
- Temperature limit:
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- USP code:
- L43, L43
- L43, L43
- L43, L43
- L43, L43
- L43, L43
- L43, L43
- L43, L43
- L43, L43
- L43
- L43
- L43, L43
- L43
- L43, L43
- L43
- L43, L43
- L43
- L43, L43