Orders LinkContactUsLinkComponent
Latex gloves, Microflex® 63-864
Latex gloves, Microflex® 63-864
Supplier:   Ansell
Latex gloves, Microflex® 63-864
Supplier:  Ansell
Certificates
Latex gloves, Microflex® 63-864
5 Options Available Below
Product Details & Documents
These disposable gloves are thicker than average Ansell latex gloves and provide chemical and liquid protection.
  • Non sterile, powder-free
  • Unique design that prevents rips and tears
  • Enhanced, textured fingertips provide a secure, dependable grip of dry and wet parts
  • Palm thickness meets and exceeds quality standards
  • Pinhole rates are lower than the standard minimum requirements for average latex gloves
Ideal for use in automotive applications, for assembly and inspection, food processing, general maintenance, painting and spraying, cleaning applications and for the handling of dry and wet parts.
Certifications: EN ISO 374-1:2016 Type B (KLT), EN ISO 374-5 VIRUS; Cat. III
Food contact approved
Documents
Specifications for Product Family
Specifications
  • Catalog No:
  • 112-4450
  • 112-4451
  • 112-4452
  • 112-4453
  • 112-4454
  • Model:
  • 63-864
  • 63-864
  • 63-864
  • 63-864
  • 63-864
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Finish:
  • Textured
  • Textured
  • Textured
  • Textured
  • Textured
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Palm thickness:
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • EN norm:
  • EN ISO 374-1:2016 Type B (KLT), EN ISO 374-5:201 VIRUS
  • EN ISO 374-1:2016 Type B (KLT), EN ISO 374-5:201 VIRUS
  • EN ISO 374-1:2016 Type B (KLT), EN ISO 374-5:201 VIRUS
  • EN ISO 374-1:2016 Type B (KLT), EN ISO 374-5:201 VIRUS
  • EN ISO 374-1:2016 Type B (KLT), EN ISO 374-5:201 VIRUS
  • Food contact approved:
  • +
  • +
  • +
  • +
  • +
Specifications
Product Family Options
Product Information
  • Glove size
    Colour
    Length
    Packed
    Availability
    Price
  • 5,5 - 6
    Natural
    245 mm
    100/box
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 27,50 €
    27,50 €
    1 * 100 items
       27,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-4450
    Model
    63-864
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Powdered / powder-free
    Powder-free
    Finish
    Textured
    Cuff style
    Beaded
    Palm thickness
    0,16 mm
    EN norm
    EN ISO 374-1:2016 Type B (KLT), EN ISO 374-5:201 VIRUS
    Food contact approved
    +
  • 6,5 - 7
    Natural
    245 mm
    100/box
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 27,50 €
    27,50 €
    1 * 100 items
       27,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-4451
    Model
    63-864
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Powdered / powder-free
    Powder-free
    Finish
    Textured
    Cuff style
    Beaded
    Palm thickness
    0,16 mm
    EN norm
    EN ISO 374-1:2016 Type B (KLT), EN ISO 374-5:201 VIRUS
    Food contact approved
    +
  • 7,5 - 8
    Natural
    245 mm
    100/box
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 27,50 €
    27,50 €
    1 * 100 items
       27,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-4452
    Model
    63-864
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Powdered / powder-free
    Powder-free
    Finish
    Textured
    Cuff style
    Beaded
    Palm thickness
    0,16 mm
    EN norm
    EN ISO 374-1:2016 Type B (KLT), EN ISO 374-5:201 VIRUS
    Food contact approved
    +
  • 8,5 - 9
    Natural
    245 mm
    100/box
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 27,50 €
    27,50 €
    1 * 100 items
       27,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-4453
    Model
    63-864
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Powdered / powder-free
    Powder-free
    Finish
    Textured
    Cuff style
    Beaded
    Palm thickness
    0,16 mm
    EN norm
    EN ISO 374-1:2016 Type B (KLT), EN ISO 374-5:201 VIRUS
    Food contact approved
    +
  • 9,5 - 10
    Natural
    245 mm
    100/box
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 27,50 €
    27,50 €
    1 * 100 items
       27,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-4454
    Model
    63-864
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Powdered / powder-free
    Powder-free
    Finish
    Textured
    Cuff style
    Beaded
    Palm thickness
    0,16 mm
    EN norm
    EN ISO 374-1:2016 Type B (KLT), EN ISO 374-5:201 VIRUS
    Food contact approved
    +