Avantor® ACE® Silica, HPLC Columns, 5 µm
Supplier: Avantor
Certificates
About this item
Avantor® ACE® Silica columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
- Ultra high purity, base deactivated silica with exceptional reproducibility
- Wide range of particle sizes (3 to 10 µm) and complementary phases
- Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.
Specifications
- Catalog No:
- ACE-127-3004
- ACE-127-3046
- ACE-127-7530
- ACE-127-1204
- ACE-127-0346
- ACE-127-7520
- ACE-127-3020
- ACE-127-2046
- ACE-127-3030
- ACE-127-3039
- ACE-127-1246
- ACE-127-1220
- ACE-127-0546
- ACE-127-2530
- ACE-127-2520
- ACE-127-0530
- ACE-127-2510
- ACE-127-0520
- ACE-127-0510
- ACE-127-0103GD
- ACE-127-0102GD
- ACE-127-1002
- ACE-127-2546
- ACE-127-0110GD
- ACE-127-0120GD
- ACE-127-1510
- ACE-127-1546
- ACE-127-1530
- ACE-127-1520
- ACE-127-2504
- ACE-127-1020
- ACE-127-1010
- ACE-127-1046
- ACE-127-2508
- ACE-127-1030
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carbon load:
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- -
- Endcapped:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Max. pressure:
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- pH range:
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- 2,0 - 7,0
- Surface area:
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- USP code:
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
Specifications
Related Information
Guard holders and accessories