Orders LinkContactUsLinkComponent
Counting chambers, Cellometer®
Counting chambers, Cellometer®
Counting chambers, Cellometer®
Counting chambers, Cellometer®
These counting chambers can be inserted into a cell counter, so that the cells are imaged directly.
  • Two independent enclosed chambers
  • Compatible with all Cellometer® cell counters
  • Cell suspension (20 μl) loaded via a a single channel pipette
Product Family Options
  • Description
  • Counting chambers with protective film
    1 * 75 items
    1 * 75 items - 898,00 €
    898,00 €
    1 * 75 items
       898,00 €
  • Counting chambers with protective film