Orders LinkContactUsLinkComponent
Qualitative filter papers, technical, smooth, grade 3 m/N
Qualitative filter papers, technical, smooth, grade 3 m/N
   Sustainable Options Available
Supplier:   Sartorius
Qualitative filter papers, technical, smooth, grade 3 m/N
Supplier:  Sartorius
   Sustainable Options Available
Certificates
Qualitative filter papers, technical, smooth, grade 3 m/N
Qualitative filter papers, technical, smooth, grade 3 m/N
12 Options Available Below
Product Details & Documents
These smooth filter papers are made of refined pulp and linters with more than 95% alpha-cellulose content. The ash content of these filter papers is between 0,1 and 0,15%. They are used for routine analyses like clarification, determination of substances, but also as discs with a centre hole for technical applications. Grades with a wet burst resistance >30 kPa are referred to as wet-strengthened and are therefore suitable for pressure or vacuum filtration.
Medium fast rate of filtration, thin, smooth, increased wet strength. Used for acids; general vacuum or pressure filtration applications and less critical analyses; also used to assay the sucrose content in cane sugar solutions with a relatively low viscosity.
Specifications for Product Family
Specifications
  • Catalog No:
  • 516-7100
  • SARTFT-3-305-110
  • 515-1591
  • 515-1592
  • SARTFT-3-305-185
  • SARTFT-3-305-240
  • SARTFT-3-305-270
  • SARTFT-4-305-125
  • SARTFT-4-305-150
  • SARTFT-4-305-185
  • SARTFT-4-305-240
  • SARTFT-4-305-385
  • Grade:
  • 3 m/N
  • 3 m/N
  • 3 m/N
  • 3 m/N
  • 3 m/N
  • 3 m/N
  • 3 m/N
  • 3 m/N
  • 3 m/N
  • 3 m/N
  • 3 m/N
  • 3 m/N
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Weight:
  • 65 g/m²
  • 65 g/m²
  • 65 g/m²
  • 65 g/m²
  • 65 g/m²
  • 65 g/m²
  • 65 g/m²
  • 65 g/m²
  • 65 g/m²
  • 65 g/m²
  • 65 g/m²
  • 65 g/m²
  • Thickness:
  • 0,14 mm
  • 0,14 mm
  • 0,14 mm
  • 0,14 mm
  • 0,14 mm
  • 0,14 mm
  • 0,14 mm
  • 0,14 mm
  • 0,14 mm
  • 0,14 mm
  • 0,14 mm
  • 0,14 mm
  • Particle retention:
  • 7 - 10 µm
  • 7 - 10 µm
  • 7 - 10 µm
  • 7 - 10 µm
  • 7 - 10 µm
  • 7 - 10 µm
  • 7 - 10 µm
  • 7 - 10 µm
  • 7 - 10 µm
  • 7 - 10 µm
  • 7 - 10 µm
  • 7 - 10 µm
  • Filtration speed:
  • 30 s
  • 30 s
  • 30 s
  • 30 s
  • 30 s
  • 30 s
  • 30 s
  • 30 s
  • 30 s
  • 30 s
  • 30 s
  • 30 s
  • Wet burst strength:
  • 30 kPa
  • 30 kPa
  • 30 kPa
  • 30 kPa
  • 30 kPa
  • 30 kPa
  • 30 kPa
  • 30 kPa
  • 30 kPa
  • 30 kPa
  • 30 kPa
  • 30 kPa
  • Environmental certification:
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
Specifications
Product Family Options
Plain filters
Product Information
  • Grade
    Size
    Availability
    Price
  • 3 m/N
    580×580 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 116,00 €
    116,00 €
    1 * 100 items
       116,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    516-7100
    Grade
    3 m/N
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    65 g/m²
    Thickness
    0,14 mm
    Particle retention
    7 - 10 µm
    Filtration speed
    30 s
    Wet burst strength
    30 kPa
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 3 m/N
    110 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 12,40 €
    12,40 €
    1 * 100 items
       12,40 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SARTFT-3-305-110
    Grade
    3 m/N
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    65 g/m²
    Thickness
    0,14 mm
    Particle retention
    7 - 10 µm
    Filtration speed
    30 s
    Wet burst strength
    30 kPa
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 3 m/N
    125 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 12,40 €
    12,40 €
    1 * 100 items
       12,40 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    515-1591
    Grade
    3 m/N
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    65 g/m²
    Thickness
    0,14 mm
    Particle retention
    7 - 10 µm
    Filtration speed
    30 s
    Wet burst strength
    30 kPa
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 3 m/N
    150 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 17,50 €
    17,50 €
    1 * 100 items
       17,50 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    515-1592
    Grade
    3 m/N
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    65 g/m²
    Thickness
    0,14 mm
    Particle retention
    7 - 10 µm
    Filtration speed
    30 s
    Wet burst strength
    30 kPa
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 3 m/N
    185 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 22,70 €
    22,70 €
    1 * 100 items
       22,70 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SARTFT-3-305-185
    Grade
    3 m/N
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    65 g/m²
    Thickness
    0,14 mm
    Particle retention
    7 - 10 µm
    Filtration speed
    30 s
    Wet burst strength
    30 kPa
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 3 m/N
    240 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 38,70 €
    38,70 €
    1 * 100 items
       38,70 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SARTFT-3-305-240
    Grade
    3 m/N
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    65 g/m²
    Thickness
    0,14 mm
    Particle retention
    7 - 10 µm
    Filtration speed
    30 s
    Wet burst strength
    30 kPa
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 3 m/N
    270 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 46,70 €
    46,70 €
    1 * 100 items
       46,70 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SARTFT-3-305-270
    Grade
    3 m/N
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    65 g/m²
    Thickness
    0,14 mm
    Particle retention
    7 - 10 µm
    Filtration speed
    30 s
    Wet burst strength
    30 kPa
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
Folded filters
Product Information
  • Grade
    Size
    Availability
    Price
  • 3 m/N
    125 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 45,60 €
    45,60 €
    1 * 100 items
       45,60 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SARTFT-4-305-125
    Grade
    3 m/N
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    65 g/m²
    Thickness
    0,14 mm
    Particle retention
    7 - 10 µm
    Filtration speed
    30 s
    Wet burst strength
    30 kPa
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 3 m/N
    150 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 45,60 €
    45,60 €
    1 * 100 items
       45,60 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SARTFT-4-305-150
    Grade
    3 m/N
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    65 g/m²
    Thickness
    0,14 mm
    Particle retention
    7 - 10 µm
    Filtration speed
    30 s
    Wet burst strength
    30 kPa
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 3 m/N
    185 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 45,60 €
    45,60 €
    1 * 100 items
       45,60 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SARTFT-4-305-185
    Grade
    3 m/N
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    65 g/m²
    Thickness
    0,14 mm
    Particle retention
    7 - 10 µm
    Filtration speed
    30 s
    Wet burst strength
    30 kPa
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 3 m/N
    240 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 77,10 €
    77,10 €
    1 * 100 items
       77,10 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SARTFT-4-305-240
    Grade
    3 m/N
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    65 g/m²
    Thickness
    0,14 mm
    Particle retention
    7 - 10 µm
    Filtration speed
    30 s
    Wet burst strength
    30 kPa
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 3 m/N
    385 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 274,00 €
    274,00 €
    1 * 100 items
       274,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SARTFT-4-305-385
    Grade
    3 m/N
    Environmentally Preferable
    Low Manufacturing Impact
    Weight
    65 g/m²
    Thickness
    0,14 mm
    Particle retention
    7 - 10 µm
    Filtration speed
    30 s
    Wet burst strength
    30 kPa
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact