Digital interval timer
Digital interval timer, 99 min, 59 s, White
HECH41977020
Digital interval timer with white casing and magnetic back.
- LCD display, 4 digits showing minutes and seconds
- Programmable up to 99 minutes and 59 seconds
- Counts down in seconds
- Alarm sounds for 30 seconds when countdown reaches zero
- Memory recall function
- Individual setting for minutes and seconds
- Colour:White
- Description:Digital interval timer, 99 min, 59 s