Anti-POLR3K Polyclonal Antibody Pair
Catalog # ABNOH00051728-AP21
Supplier: Abnova
Specifications
- Antigen name:Polymerase (RNA) III (DNA directed) Polypeptide K (12.3kDa)
- Antigen symbol:POLR3K
- Clonality:Polyclonal
- Host:Multiple
- Reactivity:Human
- Gene ID:51728
- Antigen synonyms:RPC12.5|C11-RNP3|RPC10|My010|C11|RPC11
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
Specifications
About this item
This Set comes with matched Antibody Pair to detect and quantify protein level of human POLR3K.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-POLR3K (100 µg); 2. Detection Antibody: Mouse Polyclonal Anti-POLR3K (40 µl);
Type:
Antigen: POLR3K
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human