ContactUsLinkComponent
Crystal optics, potassium bromide window, polished
Crystal optics, potassium bromide window, polished
  INCR0002C-69
Crystal optics, potassium bromide window, polished
  INCR0002C-69
  • Description:
    Polished crystal optics, KBr, 38,5×19,5×4 mm
  • Pk:
    1