Repetitive pipette, Multipette® E3 / E3x
About this item
This pipette enables precise pipetting from 1 μl to 50 ml and makes long pipetting series faster and easier. The aspirated liquid can be dispensed up to 100 times without a refill making the Multipette® the ideal tool for filling plates or large series of tubes (e.g., nucleic acid purification kits).
- To be used with Eppendorf Combitips advanced®: Automatic tip recognition for ease of use
- Defined dispensing speed increases the reproducibility of results; eight different speed levels allow the optimum speed for each application
- One button tip ejection enables one handed operation
- RFID chip contains all relevant data regarding the pipette (serial no., dispensing vol., factory adjustment, and more)
- Enlarged colour display with optimised contrast and clear arrangement of all adjustable parameters is easy to read
- Software shows all selected parameters at one glance; proven intuitive software navigation
- Display in 9 different languages
Operation modes:
E3 model: Dispensing, automatic dispensing, pipetting
E3x model: Dispensing, automatic dispensing, pipetting, aspirating, titrating, sequential dispensing, combined aspiration and dispensing mode
* Works with Windows 7/8/Vista.
Delivery information: Supplied with Combitips advanced® assortment pack (one Combitip of each size).
4 Options Available Below
Product Details & Documents
This pipette enables precise pipetting from 1 μl to 50 ml and makes long pipetting series faster and easier. The aspirated liquid can be dispensed up to 100 times without a refill making the Multipette® the ideal tool for filling plates or large series of tubes (e.g., nucleic acid purification kits).
- To be used with Eppendorf Combitips advanced®: Automatic tip recognition for ease of use
- Defined dispensing speed increases the reproducibility of results; eight different speed levels allow the optimum speed for each application
- One button tip ejection enables one handed operation
- RFID chip contains all relevant data regarding the pipette (serial no., dispensing vol., factory adjustment, and more)
- Enlarged colour display with optimised contrast and clear arrangement of all adjustable parameters is easy to read
- Software shows all selected parameters at one glance; proven intuitive software navigation
- Display in 9 different languages
Operation modes:
E3 model: Dispensing, automatic dispensing, pipetting
E3x model: Dispensing, automatic dispensing, pipetting, aspirating, titrating, sequential dispensing, combined aspiration and dispensing mode
* Works with Windows 7/8/Vista.
Delivery information: Supplied with Combitips advanced® assortment pack (one Combitip of each size).
Documents
Specifications for Product Family
Specifications
- Catalog No:
- 613-3892
- 613-3893
- 613-5189
- 613-5190
- Environmentally Preferable:
- Low Manufacturing Impact|Product Transparency and Disclosure
- Low Manufacturing Impact|Product Transparency and Disclosure
- Low Manufacturing Impact|Product Transparency and Disclosure
- Low Manufacturing Impact|Product Transparency and Disclosure
- Environmental certification:
- Renewable Energy Certificate, ISO 14001, ACT Label
- Renewable Energy Certificate, ISO 14001, ACT Label
- Renewable Energy Certificate, ISO 14001, ACT Label
- Renewable Energy Certificate, ISO 14001, ACT Label
Specifications
Product Family Options
Product Information
- AvailabilityPrice
- 613-3892Multipette® E3, with charging adapter and Combitips advanced® assortment packMultipette® E3, with charging adapter and Combitips advanced® assortment packLog in for availability1 * 1 items783,00 €Specifications:
Low Manufacturing Impact|Product Transparency and Disclosure More
Specifications
Cat. No.613-3892Environmentally PreferableLow Manufacturing Impact|Product Transparency and DisclosureEnvironmental certificationRenewable Energy Certificate, ISO 14001, ACT LabelLow Manufacturing Impact|Product Transparency and Disclosure - 613-3893Multipette® E3x, with charging adapter and Combitips advanced® assortment packMultipette® E3x, with charging adapter and Combitips advanced® assortment packLog in for availability1 * 1 items1 000,00 €Specifications:
Low Manufacturing Impact|Product Transparency and Disclosure More
Specifications
Cat. No.613-3893Environmentally PreferableLow Manufacturing Impact|Product Transparency and DisclosureEnvironmental certificationRenewable Energy Certificate, ISO 14001, ACT LabelLow Manufacturing Impact|Product Transparency and Disclosure - 613-5189Multipette® E3, with charging adapter, charging stand and Combitips advanced® assortment packMultipette® E3, with charging adapter, charging stand and Combitips advanced® assortment packLog in for availability1 * 1 items885,00 €Specifications:
Low Manufacturing Impact|Product Transparency and Disclosure More
Specifications
Cat. No.613-5189Environmentally PreferableLow Manufacturing Impact|Product Transparency and DisclosureEnvironmental certificationRenewable Energy Certificate, ISO 14001, ACT LabelLow Manufacturing Impact|Product Transparency and Disclosure - 613-5190Multipette® E3x, with charging adapter, charging stand and Combitips advanced® assortment packMultipette® E3x, with charging adapter, charging stand and Combitips advanced® assortment packLog in for availability1 * 1 items1 079,00 €Specifications:
Low Manufacturing Impact|Product Transparency and Disclosure More
Specifications
Cat. No.613-5190Environmentally PreferableLow Manufacturing Impact|Product Transparency and DisclosureEnvironmental certificationRenewable Energy Certificate, ISO 14001, ACT LabelLow Manufacturing Impact|Product Transparency and Disclosure
Complementary Products
Product Information
- Catalog #PackedVolumeColour codeSterileAvailabilityPrice
- 613-2065100 pcs. (4 bags × 25 pcs.), each box containing one adapter25 mlRed-Log in for availability1 * 100 items - 132,00 €
- 613-2066100 pcs. (4 bags × 25 pcs.), each box containing one adapter50 mlLight grey-Log in for availability1 * 100 items - 145,00 €
- 613-38571 Combitips® advanced of each size (9 total) and Adapter advanced 25 & 50 ml0,1, 0,2, 0,5, 1,0, 2,5, 5,0, 10, 25, 50 mlAssorted-Log in for availability1 * 1 KIT - 16,40 €
- 613-2059100 pcs. (4 bags × 25 pcs.)0,2 mlLight blue-Log in for availability1 * 100 items - 112,00 €
Accessories
Product Information
- Catalog #AvailabilityPrice
- Charger stand 2, for 1 Repeater® E3 or E3xCharger stand 2, for 1 Repeater® E3 or E3xEPPE3116000.040Log in for availability1 * 1 items - 286,00 €
Product Information
- Catalog #AvailabilityPrice
- Charger carousel 2, for 6 Eppendorf Xplorer® or Xplorer® plusCharger carousel 2, for 6 Eppendorf Xplorer® or Xplorer® plus613-6971Log in for availability1 * 1 items - 541,00 €
Product Information
- Catalog #AvailabilityPrice
- Charger Shell 2, for one Eppendorf Multipette® E3/E3x or Multipette® stream/Xstream, for Charger Carousel 2, with charging functionalityCharger Shell 2, for one Eppendorf Multipette® E3/E3x or Multipette® stream/Xstream, for Charger Carousel 2, with charging functionalityEPPE3116603.003Log in for availability1 * 1 items - 60,00 €
Product Information
- Catalog #AvailabilityPrice
- Power supply with plug adapters for Charger Carousel 2 and Charger Stand 2, 100 - 240 V/50 - 60 Hz (AU/EU/GB/US), with magnetic power connectorPower supply with plug adapters for Charger Carousel 2 and Charger Stand 2, 100 - 240 V/50 - 60 Hz (AU/EU/GB/US), with magnetic power connectorEPPE3116607.009Log in for availability1 * 1 items - 53,10 €
Product Information
- Catalog #AvailabilityPrice
- Li-ion-battery, rechargeable