384-well microplates, ShallowWell, standard height
Catalog # 734-2876
Supplier: Thermo Fisher Scientific
Specifications
- Recommended working volume:2 - 20 µl
- Colour:Clear
- Description:384 ShallowWell plate, non treated surface, without lid
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:NAR Certificate of Retirement
- Pk:100
- Well volume:25 µl
Specifications
About this item
These standard height plates are suitable for automation, without the need to make adjustments to robotics, plate readers or liquid handlers.
- Untreated hydrophobic polystyrene suited for growth of suspension cultures
- ANSI/SBS standard footprint