Anti-POLR3E Mouse Monoclonal Antibody [clone: 4F4]
Catalog # ABNOH00055718-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Polymerase (RNA) III (DNA directed) polypeptide E (80kDa)
- Antigen symbol:POLR3E
- Clonality:Monoclonal
- Clone:4F4
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a kappa
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:55718
- Antigen synonyms:RNA polymerase III subunit C5|RNA polymerase III 80 kDa subunit RPC5|SIN|DNA-directed RNA polymerase III subunit RPC5|DNA-directed RNA polymerase III 80 kDa polypeptide|RPC5
- Amino acid number:1 to 666
- Storage buffer:1x PBS, pH 7,4
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:POLR3E (AAH00285, 1 a.a. ~ 666 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against a full-length recombinant POLR3E.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: POLR3E
Clonality: Monoclonal
Clone: 4F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: