Anti-POLK Goat Polyclonal Antibody
Catalog # ABNOPAB11246
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Polymerase (DNA directed) kappa
- Antigen symbol:POLK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Orangutan,Chimpanzee,Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:51426
- Antigen synonyms:OTTHUMP00000020323|BETAN|BETA-N|POLL|DNA polymerase beta-2|pol lambda|polymerase (DNA-directed)|pol beta2|DNA-directed DNA polymerase lambda|POLKAPPA|polymerase (DNA directed)|DNA polymerase kappa (DNA polymerase beta-N)|POL-KAPPA|DNA polymerase lamda2|lambda|FLJ46002|RP11-529I10.1
- Amino acid number:817 to 830
- Storage buffer:20 mM KH2PO4, 150 mM NaCl, pH 7,2 (0,01% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 817-830 of human POLK.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of POLK.
Recommended Dilutions: ELISA: 1:2000-1:10000; Western Blot: 1:500-1:3000
Type: Primary
Antigen: POLK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Chimpanzee, Human, Mouse, Orangutan