Anti-CASP8 Mouse Monoclonal Antibody [clone: 90A992]
Catalog # CAYM13913-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:CASP8
- Clonality:Monoclonal
- Clone:90A992
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Rhesus monkey,Chimpanzee,Human
- Western blot:Yes
- Cross adsorption:No
- Storage buffer:PBS containing 0,05% BSA and 0,05% sodium azide
- Sequence:PVETDSEEQP
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:human caspase-8 amino acids PVETDSEEQP
- Purification:purified by Protein G chromatography
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: CASP8
Clonality: Monoclonal
Clone: 90A992
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Chimpanzee, Rhesus Monkey