Anti-TRPV3 Mouse Monoclonal Antibody [clone: S15-4]
CAYM13721-100
- Antibody type:Primary
- Antigen symbol:TRPV3
- Clonality:Monoclonal
- Clone:S15-4
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Amino acid number:458 to 474
- Storage buffer:PBS, pH 7,4, containing 50% glycerol and 0,09% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:amino acids 458-474 of rat TRPV3
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: TRPV3
Clonality: Monoclonal
Clone: S15-4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat