'Live Cell Imaging' System, JuLI™ Stage
Live cell imaging system with PC and software, JuLI™ Stage
Catalog # 734-2711
Supplier: NANOENTEK
Jump to:
Product Details & Documents
JuLI™ Stage is a fully automated, digital fluorescence imaging analyser, that directly acquires cell images from cell culture plates placed in a cell culture incubator.
- Fluorescence DAPI (ex: 390/40, em: 452/45) ; GFP (ex: 466/40, em: 525/50) ; RFP (ex: 525/50, em: 580LP)
- Three-fold fluorescence, time-lapse and image stitching functionality
- Simple to install and the equipment and software easy to operate
Applications include monitoring and analysis of cell growth, proliferation, migration and differentiation simultaneously in 1 to 384 wells; monitoring and analysis of cell expression and subcellular localisation; cell cytotoxicity and cell migration studies/scratch assays.
Documents
JuLI Stage Brochure_V0.0_E
Product Literature
JuLI Stage Live Cell Imagingsystem
Datasheet
Specifications
- Model:JuLI™ Stage
- Description:Live cell imaging system with PC and software
- Computer requirements:All-in-One Touch Screen Desktop with i5-processor, Windows 8.1 (64 bit), 8 GB RAM, 1 TB HDD, 23'' Full HD Touch Screen
- Cat. No.:734-2711
- Output:Images (JPEG, PNG, TIFF, BMP), videos (AVI), raw data (CSV)
- W×D×H:429×324×310 mm
- Stage:X/Y motorised recording in parallel several well positions from different cell culture vessels
- Camera:Monochrome CCD (Sony sensor 2/3''), 1936×1456 P (2,8 MP), 53 FPS, 14 bit with autofocus
- Weight:18 kg (without PC)
- Operating conditions:5 – 40 °C, 20 – 95 % relative humidity