Examination gloves, nitrile, PURPLE NITRILE*
Supplier: KIMBERLY CLARK
Certificates
About this item
These disposable gloves made of a highly durable nitrile polymer guard against incidental splash exposure. Ideal for medical examinations and lab applications. Also suitable for use in chemotherapy.
- Non sterile, powder-free, textured fingertips
- Excellent dexterity and tactile sensitivity
- Comfortable fit
The SmartPull* dispensing box incorporates two separate openings: One smaller opening (used when the box is full to help control dispensing to lessen waste from dropped gloves or multiple dispensing) and a second, larger opening (allows easier access to the gloves when the box is half empty).
Certifications: AQL 1,5
Frequently Bought Together