Anti-Protein Synthesis Initiation Factor 4E Binding Protein, Rabbit Polyclonal Antibody
USBIP9108-51F
- Antibody type:Primary
- Antigen name:Protein Synthesis Initiation Factor 4E Binding Protein,
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Immunochemistry: 2.5-5 ?g/ml; Western Blot: 1:1000-1:2000
Type: Primary
Antigen: Protein Synthesis Initiation Factor 4E Binding Protein,
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human