Anti-Protein Synthesis Initiation Factor 4E Binding Protein, Rabbit Polyclonal Antibody
USBIP9108-51B
- Antibody type:Primary
- Antigen name:Protein Synthesis Initiation Factor 4E Binding Protein,
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Pig,Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:Tris, pH 7,5
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Western Blot: 1:1000-1:6000
Type: Primary
Antigen: Protein Synthesis Initiation Factor 4E Binding Protein,
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Pig, Rat