Anti-HPi4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HIC1-5F10]
NOVUNBP1-18948APC
- Antibody type:Primary
- Antigen symbol:HPI4
- Clonality:Monoclonal
- Clone:HIC1-5F10
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Type: Primary
Antigen: HPi4
Clonality: monoclonal
Clone: HIC1-5F10
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human