Anti-MYC Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 9E10]
Catalog # NOVUNB600-302APC
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:v-myc avian myelocytomatosis viral oncogene homolog
- Antigen symbol:MYC
- Clonality:Monoclonal
- Clone:9E10
- Conjugation:APC (Allophycocyanin)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Western blot:Yes
- Gene ID:4609
- Antigen synonyms:MRTL|bHLHe39|c-Myc|MYCC
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: monoclonal
Clone: 90000000000
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: